Learn More
Medchemexpress LLC 3-nitrophenylhydrazine-13C6 hydrochloride | 1977535-33-3 | 98.3% | 195.56 | 13C6H8ClN3O2 | 5 MG
SDP
Catalog No. 5000262892
Encompass_Preferred
3-Nitrophenylhydrazine-13C6 hydrochloride is the 13C-labeled isotopologue of 3-nitrophenylhydrazine supplied as the hydrochloride salt. It serves as a tracer and internal standard for quantitative analytical workflows, and is commonly used after derivatization to enhance detection by NMR, GC-MS, and LC-MS.
Catalog No. 50-002-62892 Supplier Medchemexpress LLC Supplier No. HY133689AS5MG
3-Nitrophenylhydrazine-13C6 hydrochloride is the 13C-labeled isotopologue of 3-nitrophenylhydrazine supplied as the hydrochloride salt. It serves as a tracer and internal standard for quantitative analytical workflows, and is commonly used after derivatization to enhance detection by NMR, GC-MS, and LC-MS.