Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitrophthalic acid, 95%

Catalog No. AC160160025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160160025 2.5 kg Plastic bottle
AC160161000 100 g Plastic bottle
AC160165000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160160025 Supplier Thermo Scientific Chemicals Supplier No. 160160025
Add to Cart

CAS: 603-11-2 | C8H5NO6 | 211.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-11-2
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.13
InChI Key KFIRODWJCYBBHY-UHFFFAOYSA-N
Synonym 3-nitrophthalic acid, 3-nitrobenzene-1,2-dicarboxylic acid, phthalic acid, 3-nitro, 1,2-benzenedicarboxylic acid, 3-nitro, 3-nitro phthalic acid, 3-nitro-phthalic acid, 3-nitrophthalicacid, nitrophthalic acid, 3-nitrophtalic acid, m-nitrophthalic acid
PubChem CID 69043
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)C(=O)O

Specifications

Melting Point 213.0°C to 216.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 09,823
Solubility Information Solubility in water: 20g/L (25 c). Other solubilities: 1g/10mL methanol
Formula Weight 211.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Nitrophthalic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 210-030-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.