Learn More
Medchemexpress LLC HY-W012875 100mg Medchemexpress, 3-Nitropropanoic acid CAS:504-88-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW012875100MG
Encompass_Preferred
Medchemexpress, HY-W012875 100mg 3-Nitropropanoic acid CAS:504-88-1 3-Nitropropanoic acid is an irreversible inhibitor of succinate dehydrogenase . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4573