Learn More
Selleck Chemical LLC 3-Nitropropionic acid-25mg
SDP
Catalog No. 502389999
Encompass_Preferred
3-Nitropropionic acid (β-Nitropropionic Acid, 3-NP) is an irreversible inhibitor of mitochondrial respiratory Complex II succinate dehydrogenase, resulting in energy depletion through disruption of the electron transport chain.
Catalog No. 50-238-9999 Supplier Selleck Chemical LLC Supplier No. S365225MG
3-Nitropropionic acid (β-Nitropropionic Acid, 3-NP) is an irreversible inhibitor of mitochondrial respiratory Complex II succinate dehydrogenase, resulting in energy depletion through disruption of the electron transport chain.