Learn More
Abcam 3-Nitropropionic acid, Succinic dehydrogenase (comple x II) inhibitor, 50MG
Catalog No. 502705131
Encompass_Preferred
MW 119.08 Da, Purity >97%. Selective, irreversible succinic dehydrogenase (complex II) inhibitor. Induces mitochondrial citric acid cycle dysfunction, oxidative stress and cell death. Activates the Keap1-Nrf2-ARE pathway. Inhibits the MAPK and NF-κB pathways. Shows neurotoxic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5131 Supplier Abcam Supplier No. AB14558950MG
MW 119.08 Da, Purity >97%. Selective, irreversible succinic dehydrogenase (complex II) inhibitor. Induces mitochondrial citric acid cycle dysfunction, oxidative stress and cell death. Activates the Keap1-Nrf2-ARE pathway. Inhibits the MAPK and NF-κB pathways. Shows neurotoxic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement