Learn More
Medchemexpress LLC 3-Nitropyrazole | 26621-44-3 | 113.08 | 10 G
SDP
Catalog No. 5000449568
Encompass_Preferred
3-Nitropyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also a nitrogen-containing heterocyclic compound, which can be used in the synthesis of other active compounds. The nitro group of 3-Nitropyrazole readily undergoes nucleophilic substitution reactions.
  • Biochemical reagent for life science research.
  • Organic compound for life science research.
  • Nitrogen-containing heterocyclic compound.
  • Useful in the synthesis of other active compounds.
  • Nitro group readily undergoes nucleophilic substitution reactions.
Catalog No. 50-004-49568 Supplier Medchemexpress LLC Supplier No. HYW00245710G
3-Nitropyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also a nitrogen-containing heterocyclic compound, which can be used in the synthesis of other active compounds. The nitro group of 3-Nitropyrazole readily undergoes nucleophilic substitution reactions.
  • Biochemical reagent for life science research.
  • Organic compound for life science research.
  • Nitrogen-containing heterocyclic compound.
  • Useful in the synthesis of other active compounds.
  • Nitro group readily undergoes nucleophilic substitution reactions.