Learn More
Medchemexpress LLC 3-Nitropyrazole | 26621-44-3 | MFCD00159621 | 113.08 | 500 G
SDP
Catalog No. 5000449575
Encompass_Preferred
3-Nitropyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also a nitrogen-containing heterocyclic compound, which can be used in the synthesis of other active compounds. This product is for research use only and is not sold to patients.
  • Biochemical reagent for life science research
  • Nitrogen-containing heterocyclic compound
  • Can be used in the synthesis of other active compounds
  • Nitro group readily undergoes nucleophilic substitution reactions
Catalog No. 50-004-49575 Supplier Medchemexpress LLC Supplier No. HYW002457500G
3-Nitropyrazole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also a nitrogen-containing heterocyclic compound, which can be used in the synthesis of other active compounds. This product is for research use only and is not sold to patients.
  • Biochemical reagent for life science research
  • Nitrogen-containing heterocyclic compound
  • Can be used in the synthesis of other active compounds
  • Nitro group readily undergoes nucleophilic substitution reactions