Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nitropyrrole 98.0+%, TCI America™
SDP

Catalog No. N05021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05021G 1 g
N05025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05021G Supplier TCI America Supplier No. N05021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5930-94-9
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00059714
InChI Key LOJNBPNACKZWAI-UHFFFAOYSA-N
PubChem CID 145813
IUPAC Name 3-nitro-1H-pyrrole
SMILES C1=CNC=C1[N+](=O)[O-]

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Formula Weight 112.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitropyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.