Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nonanone, 98%

Catalog No. AAA1987822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1987822 100 g
AAA1987814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1987822 Supplier Thermo Scientific Chemicals Supplier No. A1987822
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 925-78-0 | C9H18O | 142.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 925-78-0
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00009541
InChI Key IYTXKIXETAELAV-UHFFFAOYSA-N
Synonym 3-nonanone, ethyl hexyl ketone, ethyl n-hexyl ketone, n-hexyl ethyl ketone, unii-00c380uhnl, 3-nonanone natural, fema no. 3440, acmc-209rga, octanone, methyl-9ci
PubChem CID 61235
IUPAC Name nonan-3-one
SMILES CCCCCCC(=O)CC

Specifications

Melting Point -8°C
Density 0.825
Boiling Point 187°C to 188°C
Flash Point 63°C (145°F)
Refractive Index 1.42
Quantity 100 g
Beilstein 1700447
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material 3-Nonanone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 213-125-2
RTECSNumber RA8227000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.