Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nonylthiophene, TCI America™
SDP

Catalog No. N05331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05331G Supplier TCI America Supplier No. N05331G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65016-63-9
Molecular Formula C13H22S
Molecular Weight (g/mol) 210.379
MDL Number MFCD00130137
InChI Key UUHSVAMCIZLNDQ-UHFFFAOYSA-N
PubChem CID 566853
IUPAC Name 3-nonylthiophene
SMILES CCCCCCCCCC1=CSC=C1

Specifications

Color Colorless
Quantity 1 g
Formula Weight 210.38
Physical Form Liquid
Chemical Name or Material 3-Nonylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.