Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nonyn-1-ol, 98%, Thermo Scientific™

Catalog No. AAA1292206
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1292206 5 g
AAA1292209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1292206 Supplier Thermo Scientific Chemicals Supplier No. A1292206

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the synthesis of 3,3,4,4- 2 H 4 -nonanol and 3,3,4,4- 2 H 4 -nonanal.

Solubility
Optimally soluble in aqueous solutions.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31333-13-8
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00002958
InChI Key TZZVRLFUTNYDEG-UHFFFAOYSA-N
Synonym 3-nonyn-1-ol, 2-hydroxyethyl pentylacetylene, 1-hydroxy-3-nonin, pubchem13073, acmc-209hlo
PubChem CID 123437
IUPAC Name non-3-yn-1-ol
SMILES CCCCCC#CCCO

Specifications

Density 0.884
Boiling Point 111°C to 112°C (20 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.457
Quantity 5 g
Beilstein 1700444
Solubility Information Optimally soluble in aqueous solutions.
Formula Weight 140.23
Percent Purity 98%
Chemical Name or Material 3-Nonyn-1-ol
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 250-573-8
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.