Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nonyn-1-ol, MP Biomedicals

Catalog No. ICN207170 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN207170 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN207170 Supplier MP Biomedicals, Inc Supplier No. 0520717005

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31333-13-8
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00002958
InChI Key TZZVRLFUTNYDEG-UHFFFAOYSA-N
Synonym 3-nonyn-1-ol, 2-hydroxyethyl pentylacetylene, 1-hydroxy-3-nonin, pubchem13073, acmc-209hlo
PubChem CID 123437
IUPAC Name non-3-yn-1-ol
SMILES CCCCCC#CCCO

Specifications

Quantity 10 g
Formula Weight 140.23
Chemical Name or Material 3-Nonyn-1-Ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.