Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Nonyn-1-ol 99.0+%, TCI America™
SDP

Catalog No. N03905ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03905ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N03905ML Supplier TCI America Supplier No. N03905ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31333-13-8
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00002958
InChI Key TZZVRLFUTNYDEG-UHFFFAOYSA-N
Synonym 3-nonyn-1-ol, 2-hydroxyethyl pentylacetylene, 1-hydroxy-3-nonin, pubchem13073, acmc-209hlo
PubChem CID 123437
IUPAC Name non-3-yn-1-ol
SMILES CCCCCC#CCCO

Specifications

Color Yellow
Boiling Point 131°C
Quantity 5 mL
Formula Weight 140.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Nonyn-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.