Learn More
Medchemexpress LLC 3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 | 163018-26-6 | C₃₀H₅₁NO₂ | 1 MG
SDP
Supplier:  Medchemexpress LLC HY152541MG
Encompass_Preferred
3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 is a Vitamin D3 derivative. This compound is intended for scientific research or pharmaceutical registration purposes only. It is a solid, white to off-white powder with a molecular weight of 457.73 and the chemical formula C₃₀H₅₁NO₂.
  • Vitamin D3 derivative
  • Intended for scientific research or pharmaceutical registration purposes
  • Soluble in DMSO at ≥ 100 mg/mL
  • Unstable in solution, fresh solutions recommended
  • Targets Vitamin D/Vitamin D receptor
Catalog No. 50-003-72422