Learn More
Medchemexpress LLC 3-O-(2-aminoethyl)-25-hydroxyvitamin D3 | 163018-26-6 | MFCD22572728 | 99.3% | 457.73 | C30H51NO2 | 10 MG
SDP
Catalog No. 5000286566
Encompass_Preferred
3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 is a synthetic derivative of 25-hydroxyvitamin D3 used as a vitamin D receptor (VDR) ligand in biomedical research. It is supplied as a high-purity solid for laboratory research; solutions are unstable and should be freshly prepared.
  • High purity (≈99.3%).
  • Acts as a ligand for the vitamin D receptor (VDR).
  • Supplied as a solid suitable for in vitro and in vivo studies.
  • Stable as a solid; unstable in solution-prepare solutions fresh.
  • Store at 4°C, protect from light, under inert atmosphere.
  • Use freshly opened DMSO to improve solubility for protocols.
Catalog No. 50-002-86566 Supplier Medchemexpress LLC Supplier No. HY1525410MG
3-O-(2-Aminoethyl)-25-hydroxyvitamin D3 is a synthetic derivative of 25-hydroxyvitamin D3 used as a vitamin D receptor (VDR) ligand in biomedical research. It is supplied as a high-purity solid for laboratory research; solutions are unstable and should be freshly prepared.
  • High purity (≈99.3%).
  • Acts as a ligand for the vitamin D receptor (VDR).
  • Supplied as a solid suitable for in vitro and in vivo studies.
  • Stable as a solid; unstable in solution-prepare solutions fresh.
  • Store at 4°C, protect from light, under inert atmosphere.
  • Use freshly opened DMSO to improve solubility for protocols.