Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-O-(2-methoxyethyl)adenosine | 303197-30-0 | 99.9% | 325.32 g·mol⁻¹ | C13H19N5O5 | 10 MG
SDP

Catalog No. 5000287633
Encompass_Preferred

3'-O-(2-methoxyethyl)adenosine is a nucleoside analogue used in biochemical and medicinal chemistry research. It is provided as a high-purity solid in small milligram quantities for laboratory experiments, including studies of nucleoside metabolism, oligonucleotide modification, and structure-activity relationships.

  • High purity (~99.9%) suitable for analytical and biochemical work.
  • Available in small research pack sizes (5 mg-200 mg).
  • Molecular formula C13H19N5O5; molecular weight 325.32 g·mol⁻¹.
  • CAS number 303197-30-0 for unambiguous identification.
  • Supplied as a stable solid for low-temperature storage and handling.

Catalog No. 50-002-87633 Supplier Medchemexpress LLC Supplier No. HY15249810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-O-(2-methoxyethyl)adenosine is a nucleoside analogue used in biochemical and medicinal chemistry research. It is provided as a high-purity solid in small milligram quantities for laboratory experiments, including studies of nucleoside metabolism, oligonucleotide modification, and structure-activity relationships.

  • High purity (~99.9%) suitable for analytical and biochemical work.
  • Available in small research pack sizes (5 mg-200 mg).
  • Molecular formula C13H19N5O5; molecular weight 325.32 g·mol⁻¹.
  • CAS number 303197-30-0 for unambiguous identification.
  • Supplied as a stable solid for low-temperature storage and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.