Learn More
Medchemexpress LLC 3'-O-(2-Methoxyethyl)adenosine | 303197-30-0 | C13H19N5O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15249850MG
Encompass_Preferred
3'-O-(2-Methoxyethyl)adenosine is an adenosine analogue that functions as a smooth muscle vasodilator and has been shown to inhibit cancer progression. This product is for research use only.
  • Adenosine analogue
  • Smooth muscle vasodilator
  • Inhibits cancer progression
  • For research use only
Catalog No. 50-003-73061