Learn More
Medchemexpress LLC 3'-O-(2-Methoxyethyl)guanosine | 256224-03-0 | C13H19N5O6 | 250 MG
SDP
Catalog No. 5000374292
Encompass_Preferred
3'-O-(2-Methoxyethyl)guanosine is a guanosine analogue. It exhibits immunostimulatory activity, and in some animal models, it induces type I interferons, leading to antiviral effects. Its functional activity is dependent on the activation of Toll-like receptor 7 (TLR7).
  • Guanosine analogue
  • Immunostimulatory activity
  • Induces type I interferons
  • Produces antiviral effects
  • Functional activity dependent on activation of Toll-like receptor 7 (TLR7)
Catalog No. 50-003-74292 Supplier Medchemexpress LLC Supplier No. 250MGHY152493
3'-O-(2-Methoxyethyl)guanosine is a guanosine analogue. It exhibits immunostimulatory activity, and in some animal models, it induces type I interferons, leading to antiviral effects. Its functional activity is dependent on the activation of Toll-like receptor 7 (TLR7).
  • Guanosine analogue
  • Immunostimulatory activity
  • Induces type I interferons
  • Produces antiviral effects
  • Functional activity dependent on activation of Toll-like receptor 7 (TLR7)