Learn More
Medchemexpress LLC 3'-O-(2-Methoxyethyl)guanosine | 256224-03-0 | C13H19N5O6 | 50 MG
SDP
Catalog No. 5000372413
Encompass_Preferred
3'-O-(2-Methoxyethyl)guanosine is a guanosine analogue. Some guanosine analogs have immunostimulatory activity. In some animal models, they also induce type I interferons, producing antiviral effects. Studies have shown that the functional activity of guanosine analogs is dependent on the activation of Toll-like receptor 7 (TLR7).
  • Guanosine analogue
  • Immunostimulatory activity
  • Induces type I interferons, producing antiviral effects
  • Functional activity dependent on activation of Toll-like receptor 7 (TLR7)
Catalog No. 50-003-72413 Supplier Medchemexpress LLC Supplier No. HY15249350MG
3'-O-(2-Methoxyethyl)guanosine is a guanosine analogue. Some guanosine analogs have immunostimulatory activity. In some animal models, they also induce type I interferons, producing antiviral effects. Studies have shown that the functional activity of guanosine analogs is dependent on the activation of Toll-like receptor 7 (TLR7).
  • Guanosine analogue
  • Immunostimulatory activity
  • Induces type I interferons, producing antiviral effects
  • Functional activity dependent on activation of Toll-like receptor 7 (TLR7)