Learn More
Medchemexpress LLC HY-N2994 1mg Medchemexpress, 3-O-Acetyl-16α-hydroxytrametenolic acid CAS:168293-13-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN29941MG
Encompass_Preferred
Medchemexpress, HY-N2994 1mg 3-O-Acetyl-16α-hydroxytrametenolic acid CAS:168293-13-8 3-O-Acetyl-16α-hydroxytrametenolic acid (Compound 3) is a triterpene found in Poria cocos, with anti-oxidant and anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0408