Learn More
TARGETMOL CHEMICALS INC 3-O-Acetyloleanolic acid 5MG
SDP
Catalog No. 502879304
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 3-O-acetyloleanolic acid dose-dependently inhibited the viability of HCT-116 cells. Apoptosis was characterized by detection of cell surface annexin V and sub-G1 apoptotic cell populations. Purity 99.2%
Catalog No. 50-287-9304 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S13965MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 3-O-acetyloleanolic acid dose-dependently inhibited the viability of HCT-116 cells. Apoptosis was characterized by detection of cell surface annexin V and sub-G1 apoptotic cell populations. Purity 99.2%