Learn More
AdipoGen 3-O-Caffeoyl-betulin
SDP
Supplier:  AdipoGen BVT0393M001
Encompass_Preferred
Chemical. CAS 89130-86-9. Formula C39H56O5. MW 604.9. Isolated from birch bark Betula alba. Anti-proliferative. Shows anti-melanoma activity. Sun-block ingredient for cosmetics. Prevents sun burning, premature aging and skin cancer. Antibacterial.
Catalog No. 50-256-1954