Learn More
Cayman Chemical 3-O-ethyl-L-Ascorbic Acid, Purity: ≥98%, CAS: 86404-04-8, 25 g
SDP
Catalog No. 502950625
Encompass_Preferred
A tyrosinase inhibitor (IC50s = 16.65 and 29.69 mM for the human and mushroom enzymes, respectively); scavenges DPPH radicals (IC50 = 0.032 g/L)
Catalog No. 50-295-0625 Supplier Cayman Chemical Supplier No. 3413925 G
A tyrosinase inhibitor (IC50s = 16.65 and 29.69 mM for the human and mushroom enzymes, respectively); scavenges DPPH radicals (IC50 = 0.032 g/L)