Learn More
Medchemexpress LLC 3-O-Ethyl-L-ascorbic acid | 86404-04-8 | 204.18 | 1 G
SDP
Catalog No. 5000445749
Encompass_Preferred
3-O-Ethyl-L-ascorbic acid is a stable derivative of vitamin C and a cosmetic tyrosinase inhibitor. It possesses antioxidant, anti-aging, and skin-whitening activities, making it suitable for use in cosmetic studies. This product is for research use only.
  • Stable derivative of vitamin C
  • Cosmetic tyrosinase inhibitor
  • Antioxidant, anti-aging, and skin-whitening activities
  • Often used in cosmetic studies
Catalog No. 50-004-45749 Supplier Medchemexpress LLC Supplier No. HYW0036071G
3-O-Ethyl-L-ascorbic acid is a stable derivative of vitamin C and a cosmetic tyrosinase inhibitor. It possesses antioxidant, anti-aging, and skin-whitening activities, making it suitable for use in cosmetic studies. This product is for research use only.
  • Stable derivative of vitamin C
  • Cosmetic tyrosinase inhibitor
  • Antioxidant, anti-aging, and skin-whitening activities
  • Often used in cosmetic studies