Learn More
Medchemexpress LLC 3-O-Ethyl-L-ascorbic acid | 86404-04-8 | 204.18 | 5 G
SDP
Catalog No. 5000449110
Encompass_Preferred
This product is a stable derivative of vitamin C and functions as a cosmetic tyrosinase inhibitor. It demonstrates antioxidant, anti-aging, and skin-whitening properties. It is often employed in cosmetic research, with a cosmetic serum containing this derivative showing good biocompatibility, excellent photoprotection, promotion of collagen synthesis, and reduction of melanin content, thereby assisting in preventing skin aging and pigmentation.
  • Stable derivative of vitamin C
  • Functions as a cosmetic tyrosinase inhibitor
  • Demonstrates antioxidant, anti-aging, and skin-whitening properties
  • Often employed in cosmetic research
  • Promotes collagen synthesis
  • Reduces melanin content
  • Assists in preventing skin aging and pigmentation
  • Exhibits good biocompatibility
  • Provides excellent photoprotection
Catalog No. 50-004-49110 Supplier Medchemexpress LLC Supplier No. HYW0036075G
This product is a stable derivative of vitamin C and functions as a cosmetic tyrosinase inhibitor. It demonstrates antioxidant, anti-aging, and skin-whitening properties. It is often employed in cosmetic research, with a cosmetic serum containing this derivative showing good biocompatibility, excellent photoprotection, promotion of collagen synthesis, and reduction of melanin content, thereby assisting in preventing skin aging and pigmentation.
  • Stable derivative of vitamin C
  • Functions as a cosmetic tyrosinase inhibitor
  • Demonstrates antioxidant, anti-aging, and skin-whitening properties
  • Often employed in cosmetic research
  • Promotes collagen synthesis
  • Reduces melanin content
  • Assists in preventing skin aging and pigmentation
  • Exhibits good biocompatibility
  • Provides excellent photoprotection