Learn More
Medchemexpress LLC 3-O-Ethyl-L-ascorbic acid | 86404-04-8 | MFCD09261382 | 204.18 | 1 ML
SDP
Catalog No. 5000447458
Encompass_Preferred
3-O-Ethyl-L-ascorbic acid is a chemical compound with a molecular weight of 204.18. It boasts a high purity of 99.9% and is supplied as a 10 mM solution in 1 mL of water.
  • For powder storage: -20 °C for 3 years, 4 °C for 2 years
  • For solutions: -80 °C for 6 months, -20 °C for 1 month
  • Detailed datasheet, SDS, and handling instructions available
Catalog No. 50-004-47458 Supplier Medchemexpress LLC Supplier No. HYW00360710MM/1ML
3-O-Ethyl-L-ascorbic acid is a chemical compound with a molecular weight of 204.18. It boasts a high purity of 99.9% and is supplied as a 10 mM solution in 1 mL of water.
  • For powder storage: -20 °C for 3 years, 4 °C for 2 years
  • For solutions: -80 °C for 6 months, -20 °C for 1 month
  • Detailed datasheet, SDS, and handling instructions available