Learn More
Medchemexpress LLC 3'-O-Me-G(iBu)-2'-phosphoramidite | 179479-04-0 | C45H56N7O9P | 50 MG
SDP
Catalog No. 5000376688
Encompass_Preferred
3'-O-Me-G(iBu)-2'-phosphoramidite is a guanosine analog that exhibits immunostimulatory activity. In some animal models, it induces type I interferons, leading to antiviral effects. Its functional activity depends on the activation of Toll-like receptor 7 (TLR7).
  • Guanosine analog
  • Exhibits immunostimulatory activity
  • Induces type I interferons in animal models
  • Leads to antiviral effects
  • Functional activity dependent on activation of Toll-like receptor 7 (TLR7)
Catalog No. 50-003-76688 Supplier Medchemexpress LLC Supplier No. HY15448450MG
3'-O-Me-G(iBu)-2'-phosphoramidite is a guanosine analog that exhibits immunostimulatory activity. In some animal models, it induces type I interferons, leading to antiviral effects. Its functional activity depends on the activation of Toll-like receptor 7 (TLR7).
  • Guanosine analog
  • Exhibits immunostimulatory activity
  • Induces type I interferons in animal models
  • Leads to antiviral effects
  • Functional activity dependent on activation of Toll-like receptor 7 (TLR7)