Learn More
New England Biolabs, Inc. 3'-O-Me-m7G(5')ppp(5')G RNA Cap Structure Analog – 5 umol
SDP
Catalog No. 50994958
Encompass
Blocking of the 3' -hydroxyl of m7G with 3' -0-Me assures that the capped transcripts are homogeneous. The 3' - hydroxyl of the non-methylated G is the only 3' - hydroxyl available for initiation. The 5' terminal m7G cap present on most eukaryotic mRNAs promotes translation in vitro at the initiation level. For most RNAs, elimination of the cap structure causes a loss of stability, especially against exonuclease degradation, and a decrease in the formation of the initiation complex of mRNAs for protein synthesis. Certain prokaryotic mRNAs containing a 5 terminal cap structure are translated as efficiently as or more efficiently than eukaryotic mRNAs in a eukaryotic cell-free protein synthesizing system. Also a cap requirement has been observed for splicing eukaryotic substrate RNAs. A method using E. coli RNA Polymerase primed with m7G(5' )ppp(5' )G or m7G(5' )ppp(5' )A for an efficient in vitro synthesis of capped RNAs has been developed by Contreas.
Catalog No. 50-994-958 Supplier New England Biolabs, Inc. Supplier No. S1411L
Blocking of the 3' -hydroxyl of m7G with 3' -0-Me assures that the capped transcripts are homogeneous. The 3' - hydroxyl of the non-methylated G is the only 3' - hydroxyl available for initiation. The 5' terminal m7G cap present on most eukaryotic mRNAs promotes translation in vitro at the initiation level. For most RNAs, elimination of the cap structure causes a loss of stability, especially against exonuclease degradation, and a decrease in the formation of the initiation complex of mRNAs for protein synthesis. Certain prokaryotic mRNAs containing a 5 terminal cap structure are translated as efficiently as or more efficiently than eukaryotic mRNAs in a eukaryotic cell-free protein synthesizing system. Also a cap requirement has been observed for splicing eukaryotic substrate RNAs. A method using E. coli RNA Polymerase primed with m7G(5' )ppp(5' )G or m7G(5' )ppp(5' )A for an efficient in vitro synthesis of capped RNAs has been developed by Contreas.