Learn More
Medchemexpress LLC 3-O-Methyl-D-chiro-inositol | 10284-63-6 | 194.18 | 1 ML
SDP
Catalog No. 5000388811
Encompass_Preferred
D-pinitol is a natural compound found in various plants, including Pinaceae and Leguminosae. It is recognized for its hypoglycemic activity and its protective effects on the cardiovascular system. Additionally, D-pinitol exhibits antiviral and larvicidal properties.
  • Natural compound found in plants.
  • Exerts hypoglycemic activity.
  • Provides protective effects in the cardiovascular system.
  • Possesses antiviral activity.
  • Exhibits larvicidal activity.
  • Promotes apoptosis in cells.
Catalog No. 50-003-88811 Supplier Medchemexpress LLC Supplier No. HYN065510MM/1ML
D-pinitol is a natural compound found in various plants, including Pinaceae and Leguminosae. It is recognized for its hypoglycemic activity and its protective effects on the cardiovascular system. Additionally, D-pinitol exhibits antiviral and larvicidal properties.
  • Natural compound found in plants.
  • Exerts hypoglycemic activity.
  • Provides protective effects in the cardiovascular system.
  • Possesses antiviral activity.
  • Exhibits larvicidal activity.
  • Promotes apoptosis in cells.