Learn More
Medchemexpress LLC 3-O-methyl-D-glucopyranose | 13224-94-7 | MFCD00006608 | 99.9% | 194.18 g/mol | C7H14O6 | 250 MG
SDP
Catalog No. 5000283702
Encompass_Preferred
3-O-Methyl-D-glucopyranose is a methylated glucose derivative supplied as a biochemical reagent for laboratory research. It acts as a non-metabolizable glucose analog useful in physiological studies of transport and uptake, and as a tracer or reference in analytical methods. The material is provided as a high-purity dry powder in research-scale pack sizes.
  • Methylated glucose derivative used as a biochemical reagent.
  • Non-metabolizable glucose analog for transport and uptake studies.
  • Suitable as a tracer or internal standard in analytical methods.
  • High purity (99.93%) for reproducible results.
  • Supplied as a stable dry powder in laboratory pack sizes.
Catalog No. 50-002-83702 Supplier Medchemexpress LLC Supplier No. HYW145482250MG
3-O-Methyl-D-glucopyranose is a methylated glucose derivative supplied as a biochemical reagent for laboratory research. It acts as a non-metabolizable glucose analog useful in physiological studies of transport and uptake, and as a tracer or reference in analytical methods. The material is provided as a high-purity dry powder in research-scale pack sizes.
  • Methylated glucose derivative used as a biochemical reagent.
  • Non-metabolizable glucose analog for transport and uptake studies.
  • Suitable as a tracer or internal standard in analytical methods.
  • High purity (99.93%) for reproducible results.
  • Supplied as a stable dry powder in laboratory pack sizes.