Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-O-Methyl-D-glucopyranose | 13224-94-7 | MFCD00063266 | 99.9% | 194.18 g/mol | C7H14O6 | 1 G
SDP

Catalog No. 5000282902
Encompass_Preferred

3-O-Methyl-D-glucopyranose is a methylated glucose derivative supplied as a biochemical reagent for laboratory research. It is commonly used as a non-metabolizable glucose analog and substrate in transport, uptake, and metabolic studies, provided as a white to off-white powder for experimental assays.

  • High purity: 99.93%.
  • Molecular formula: C7H14O6.
  • Molecular weight: 194.18 g/mol.
  • Non-metabolizable glucose analog for transport and metabolic assays.
  • Available in multiple laboratory sizes, including 1 G.
  • Includes structural identifiers suitable for computational use (SMILES, InChI).

Catalog No. 50-002-82902 Supplier Medchemexpress LLC Supplier No. HYW1454821G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-O-Methyl-D-glucopyranose is a methylated glucose derivative supplied as a biochemical reagent for laboratory research. It is commonly used as a non-metabolizable glucose analog and substrate in transport, uptake, and metabolic studies, provided as a white to off-white powder for experimental assays.

  • High purity: 99.93%.
  • Molecular formula: C7H14O6.
  • Molecular weight: 194.18 g/mol.
  • Non-metabolizable glucose analog for transport and metabolic assays.
  • Available in multiple laboratory sizes, including 1 G.
  • Includes structural identifiers suitable for computational use (SMILES, InChI).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.