Learn More
Medchemexpress LLC 3-O-methyl-D-glucopyranose | 13224-94-7 | MFCD00063266 | 99.9% | 194.18 g/mol | C7H14O6 | 5 G
SDP
Catalog No. 5000283083
Encompass_Preferred
3-O-Methyl-D-glucopyranose is a biochemical reagent used in life-science research as a non-metabolizable glucose analog for studies of glucose transport and intracellular uptake. Supplied as a white to off-white powder intended for laboratory research use only.
  • High purity: 99.9%.
  • Non-metabolizable glucose analog for transport studies.
  • White to off-white solid, suitable for assay preparation.
  • Molecular weight 194.18 g/mol and CAS number 13224-94-7 for unambiguous identification.
  • Stable under recommended storage conditions to preserve integrity.
Catalog No. 50-002-83083 Supplier Medchemexpress LLC Supplier No. HYW1454825G
3-O-Methyl-D-glucopyranose is a biochemical reagent used in life-science research as a non-metabolizable glucose analog for studies of glucose transport and intracellular uptake. Supplied as a white to off-white powder intended for laboratory research use only.
  • High purity: 99.9%.
  • Non-metabolizable glucose analog for transport studies.
  • White to off-white solid, suitable for assay preparation.
  • Molecular weight 194.18 g/mol and CAS number 13224-94-7 for unambiguous identification.
  • Stable under recommended storage conditions to preserve integrity.