Learn More
Medchemexpress LLC 3-O-methyl-D-glucose-13C | 478529-34-9 | 99.0% | (13C)C6H14O6 | 10MG
SDP
Catalog No. 5000261599
Encompass_Preferred
3-O-Methyl-D-glucose-13C is a 13C-labeled stable isotope of 3-O-methyl-D-glucose used as a high-purity tracer and internal standard in quantitative analytical workflows, including NMR, GC-MS, and LC-MS. Supplied as a solid suitable for weighing and preparing standard solutions, it supports tracer experiments and method development in biochemical and analytical laboratories.
  • High isotopic labeling for sensitive tracer and quantitation studies.
  • Purity 99.0% for reliable analytical performance.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in multiple small-scale sizes for method development.
  • Stable solid form convenient for weighing and dissolution.
Catalog No. 50-002-61599 Supplier Medchemexpress LLC Supplier No. HY146993S10MG
3-O-Methyl-D-glucose-13C is a 13C-labeled stable isotope of 3-O-methyl-D-glucose used as a high-purity tracer and internal standard in quantitative analytical workflows, including NMR, GC-MS, and LC-MS. Supplied as a solid suitable for weighing and preparing standard solutions, it supports tracer experiments and method development in biochemical and analytical laboratories.
  • High isotopic labeling for sensitive tracer and quantitation studies.
  • Purity 99.0% for reliable analytical performance.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in multiple small-scale sizes for method development.
  • Stable solid form convenient for weighing and dissolution.