Learn More
Medchemexpress LLC 3-O-Methyl-D-glucose-13C | 478529-34-9 | >99.0% | 195.18 | 100 MG
SDP
Catalog No. 5000360423
Encompass_Preferred
3-O-Methyl-D-glucose-13C is the 13C labeled 3-O-Methyl-D-glucose. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-60423 Supplier Medchemexpress LLC Supplier No. HY146993S100MG
3-O-Methyl-D-glucose-13C is the 13C labeled 3-O-Methyl-D-glucose. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS