Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-O-Methyl-DL-DOPA | 7636-26-2 | 100.0% | 211.21 | 50 MG
SDP

Catalog No. 5000435844
Encompass_Preferred

3-O-Methyl-DL-DOPA is an endogenous metabolite present in Cerebrospinal Fluid. It can be used for the research of Epilepsy, Purine Nucleoside Phosphorylase Deficiency and Aromatic L Amino Acid Decarboxylase Deficiency.

  • Purity: 99.94%
  • Appearance: solid
  • Color: off-white to gray
  • CAS No.: 7636-26-2
  • Molecular weight: 211.21
  • Formula: C10H13NO4
  • Initial source: endogenous metabolite
  • Solubility: 5 mg/mL in H2O
  • Storage: 4°C, sealed storage, away from moisture
  • Used for research of epilepsy, purine nucleoside phosphorylase deficiency and aromatic l amino acid decarboxylase deficiency

Catalog No. 50-004-35844 Supplier Medchemexpress LLC Supplier No. HY113468R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-O-Methyl-DL-DOPA is an endogenous metabolite present in Cerebrospinal Fluid. It can be used for the research of Epilepsy, Purine Nucleoside Phosphorylase Deficiency and Aromatic L Amino Acid Decarboxylase Deficiency.

  • Purity: 99.94%
  • Appearance: solid
  • Color: off-white to gray
  • CAS No.: 7636-26-2
  • Molecular weight: 211.21
  • Formula: C10H13NO4
  • Initial source: endogenous metabolite
  • Solubility: 5 mg/mL in H2O
  • Storage: 4°C, sealed storage, away from moisture
  • Used for research of epilepsy, purine nucleoside phosphorylase deficiency and aromatic l amino acid decarboxylase deficiency

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.