Learn More
Cayman Chemical 3-O-methyl-L-DOPA (hydrate), Purity: ≥98%, CAS: 200630-46-2, 10 mg
SDP
Catalog No. 502984213
Encompass_Preferred
An active metabolite of L-DOPA; inhibits L-DOPA-induced neuroprotection in rat primary mesencephalic dopaminergic neurons at 10 and 100 µM
Catalog No. 50-298-4213 Supplier Cayman Chemical Supplier No. 2073710 MG
An active metabolite of L-DOPA; inhibits L-DOPA-induced neuroprotection in rat primary mesencephalic dopaminergic neurons at 10 and 100 µM