Learn More
Medchemexpress LLC 3'-O-Methylbatatasin III | 101330-69-2 | 10 MG
SDP
Catalog No. 5000417343
Encompass_Preferred
3'-O-Methylbatatasin III exhibits spasmolytic activity and demonstrates anti-inflammatory activity in human neutrophils by inhibiting FMLP/cytochalasin B-induced superoxide anion generation and elastase release. It is intended for research use only.
  • Spasmolytic activity
  • Anti-inflammatory activity
  • Purity of 99.92%
Catalog No. 50-004-17343 Supplier Medchemexpress LLC Supplier No. HYN817510MG
3'-O-Methylbatatasin III exhibits spasmolytic activity and demonstrates anti-inflammatory activity in human neutrophils by inhibiting FMLP/cytochalasin B-induced superoxide anion generation and elastase release. It is intended for research use only.
  • Spasmolytic activity
  • Anti-inflammatory activity
  • Purity of 99.92%