Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-O-Methylbatatasin III | 101330-69-2 | 5 MG
SDP

Catalog No. 5000417370
Encompass_Preferred

3'-O-Methylbatatasin III is a chemical compound that shows spasmolytic activity. With a molecular weight of 258.31 and a molecular formula of C16H18O3, it appears as a white to off-white solid with 99.92% purity. This compound is derived from plants, specifically Orchidaceae, Bletilla striata (Thunb. ex Murray) Rchb. f., and is soluble in DMSO.

  • Shows spasmolytic activity
  • Molecular weight: 258.31
  • Molecular formula: C16H18O3
  • Appearance: Solid, white to off-white
  • Purity: 99.92%
  • Derived from plants
  • Soluble in DMSO: ≥ 100 mg/mL

Catalog No. 50-004-17370 Supplier Medchemexpress LLC Supplier No. HYN81755MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-O-Methylbatatasin III is a chemical compound that shows spasmolytic activity. With a molecular weight of 258.31 and a molecular formula of C16H18O3, it appears as a white to off-white solid with 99.92% purity. This compound is derived from plants, specifically Orchidaceae, Bletilla striata (Thunb. ex Murray) Rchb. f., and is soluble in DMSO.

  • Shows spasmolytic activity
  • Molecular weight: 258.31
  • Molecular formula: C16H18O3
  • Appearance: Solid, white to off-white
  • Purity: 99.92%
  • Derived from plants
  • Soluble in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.