Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3'-O-methylcytidine | 20594-00-7 | 98.0% | 257.24 g/mol | C10H15N3O5 | 1 ML
SDP

Catalog No. 5000284696
Encompass_Preferred

3'-O-Methylcytidine is a cytidine nucleoside analog supplied for research use. It is offered both as a solid and as pre-made stock solutions for biochemical and nucleic acid studies, and is characterized by defined molecular properties and recommended storage conditions.

  • Molecular formula C10H15N3O5 and molecular weight 257.24 g/mol.
  • Purity: 98.0% as reported by the manufacturer.
  • Soluble in DMSO at ≥100 mg/mL (≈388.7 mM), suitable for stock solution preparation.
  • Available in small solid quantities and common stock concentrations for immediate use.
  • Store solid protected from light at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended for research use only; follow appropriate safety and handling procedures.

Catalog No. 50-002-84696 Supplier Medchemexpress LLC Supplier No. HYW57827510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3'-O-Methylcytidine is a cytidine nucleoside analog supplied for research use. It is offered both as a solid and as pre-made stock solutions for biochemical and nucleic acid studies, and is characterized by defined molecular properties and recommended storage conditions.

  • Molecular formula C10H15N3O5 and molecular weight 257.24 g/mol.
  • Purity: 98.0% as reported by the manufacturer.
  • Soluble in DMSO at ≥100 mg/mL (≈388.7 mM), suitable for stock solution preparation.
  • Available in small solid quantities and common stock concentrations for immediate use.
  • Store solid protected from light at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended for research use only; follow appropriate safety and handling procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.