Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3′-O-methylcytidine | 20594-00-7 | 98.0% | 257.24 g/mol | C10H15N3O5 | 250 MG
SDP

Catalog No. 5000284655
Encompass_Preferred

3′-O-Methylcytidine is a cytidine analog provided as a solid research reagent. It is used as a nucleoside antimetabolite in studies of nucleic acid chemistry and RNA/DNA modification pathways, and is supplied with characterization data appropriate for research applications.

  • High purity (98.0%).
  • Molecular formula C10H15N3O5 and molecular weight 257.24 g/mol.
  • Provided as a solid, white to off-white.
  • Suitable for nucleic acid and methylation studies.
  • Available in a 250 mg package.
  • Storage: store at 4°C and protect from light; in solvent store at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-84655 Supplier Medchemexpress LLC Supplier No. HYW578275250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3′-O-Methylcytidine is a cytidine analog provided as a solid research reagent. It is used as a nucleoside antimetabolite in studies of nucleic acid chemistry and RNA/DNA modification pathways, and is supplied with characterization data appropriate for research applications.

  • High purity (98.0%).
  • Molecular formula C10H15N3O5 and molecular weight 257.24 g/mol.
  • Provided as a solid, white to off-white.
  • Suitable for nucleic acid and methylation studies.
  • Available in a 250 mg package.
  • Storage: store at 4°C and protect from light; in solvent store at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.