Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3′-O-methylcytidine | 20594-00-7 | MFCD00057449 | 98.0% | 257.24 g·mol⁻¹ | C10H15N3O5 | 500 MG
SDP

Catalog No. 5000284292
Encompass_Preferred

3′-O-Methylcytidine is a cytidine nucleoside analog supplied for laboratory research. It is used to study nucleic acid metabolism and DNA methyltransferase activity. The compound has CAS 20594-00-7, chemical formula C10H15N3O5, molecular weight 257.24 g·mol⁻¹, and is typically provided as a solid for experimental use.

  • Cytidine analog used in studies of nucleic acid metabolism and methylation.
  • Purity 98.0%.
  • Available as a solid; common pack size 500 MG.
  • Storage: solid at 4°C, protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only.

Catalog No. 50-002-84292 Supplier Medchemexpress LLC Supplier No. HYW578275500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3′-O-Methylcytidine is a cytidine nucleoside analog supplied for laboratory research. It is used to study nucleic acid metabolism and DNA methyltransferase activity. The compound has CAS 20594-00-7, chemical formula C10H15N3O5, molecular weight 257.24 g·mol⁻¹, and is typically provided as a solid for experimental use.

  • Cytidine analog used in studies of nucleic acid metabolism and methylation.
  • Purity 98.0%.
  • Available as a solid; common pack size 500 MG.
  • Storage: solid at 4°C, protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.