Learn More
Medchemexpress LLC 3-O-methyldopa (3-methoxy-L-tyrosine) | 300-48-1 | MFCD00037205; MFCD07784093 | 99.9% | 211.21 g/mol | C10H13NO4 | 5 MG
SDP
Catalog No. 5000263915
Encompass_Preferred
3-O-methyldopa (3-methoxy-L-tyrosine) is an analytical standard and a metabolic derivative of L-DOPA used for research and analytical applications. It can cross the blood-brain barrier and is used in studies of neuropharmacology, Parkinson's disease, and related nervous system disorders. Certificates of analysis are available for batch verification.
  • Analytical standard suitable for quantitative analysis.
  • Metabolite of L-DOPA that crosses the blood-brain barrier.
  • Applicable to neuropharmacology and Parkinson's disease research.
  • Supplied as solid and ready-to-use solution formats.
  • High purity with batch-specific certificate of analysis available.
Catalog No. 50-002-63915 Supplier Medchemexpress LLC Supplier No. HY113468AR5MG
3-O-methyldopa (3-methoxy-L-tyrosine) is an analytical standard and a metabolic derivative of L-DOPA used for research and analytical applications. It can cross the blood-brain barrier and is used in studies of neuropharmacology, Parkinson's disease, and related nervous system disorders. Certificates of analysis are available for batch verification.
  • Analytical standard suitable for quantitative analysis.
  • Metabolite of L-DOPA that crosses the blood-brain barrier.
  • Applicable to neuropharmacology and Parkinson's disease research.
  • Supplied as solid and ready-to-use solution formats.
  • High purity with batch-specific certificate of analysis available.