Learn More
Medchemexpress LLC HY-113468A 5mg , 3-O-Methyldopa CAS:300-48-1 Purity:>98%
SDP
Catalog No. 502260559
Encompass_Preferred
HY-113468A 5mg 3-O-Methyldopa CAS: 300-48-1 3-O-Methyldopa (3-Methoxy-L-tyrosine) is a metabolite of L-DOPA which is formed by catechol-O-methyltransferase (COMT). 3-O-Methyldopa competitively inhibits the pharmacodynamics of l-DOPA and dopamine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0559 Supplier Medchemexpress LLC Supplier No. HY113468A5MG
Add to Cart
Add to Cart
HY-113468A 5mg 3-O-Methyldopa CAS: 300-48-1 3-O-Methyldopa (3-Methoxy-L-tyrosine) is a metabolite of L-DOPA which is formed by catechol-O-methyltransferase (COMT). 3-O-Methyldopa competitively inhibits the pharmacodynamics of l-DOPA and dopamine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.