Learn More
Medchemexpress LLC HY-113468AS 1mg Medchemexpress, 3-O-Methyldopa D3 CAS:586954-09-8 Purity:>98%
SDP
Catalog No. 502080859
Encompass_Preferred
Medchemexpress, HY-113468AS 1mg 3-O-Methyldopa D3 CAS:586954-09-8 3-O-Methyldopa D3 (3-Methoxy-L-tyrosine D3) is deuterium labeled 3-O-Methyldopa. 3-O-Methyldopa is a metabolite of L-DOPA which is formed by catechol-O-methyltransferase (COMT). 3-O-Methyldopa competitively inhibits the pharmacodynamics of l-DOPA and dopamine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0859 Supplier Medchemexpress LLC Supplier No. HY113468AS1MG
Medchemexpress, HY-113468AS 1mg 3-O-Methyldopa D3 CAS:586954-09-8 3-O-Methyldopa D3 (3-Methoxy-L-tyrosine D3) is deuterium labeled 3-O-Methyldopa. 3-O-Methyldopa is a metabolite of L-DOPA which is formed by catechol-O-methyltransferase (COMT). 3-O-Methyldopa competitively inhibits the pharmacodynamics of l-DOPA and dopamine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.