Learn More
Medchemexpress LLC 3-O-methyldopa-d3 hydrate | MFCD11983476 | 232.25 | C10H12D3NO5 | 5 MG
SDP
Catalog No. 5000264244
Encompass_Preferred
3-O-Methyldopa-d3 hydrate is the deuterium-labeled form of 3-O-methyldopa supplied as a white to off-white solid hydrate. It is designed for use as an internal standard or tracer in quantitative analyses and for research on dopamine metabolism and related neurological disorders.
  • Deuterium-labeled internal standard for LC-MS, GC-MS, and NMR.
  • Suitable for quantitative tracer and assay development.
  • Solid hydrate form with stable isotopic labeling.
  • Useful in studies of dopamine metabolism and neurological disease models.
  • Supplied in small research-scale quantities for analytical applications.
Catalog No. 50-002-64244 Supplier Medchemexpress LLC Supplier No. HY113468AS15MG
3-O-Methyldopa-d3 hydrate is the deuterium-labeled form of 3-O-methyldopa supplied as a white to off-white solid hydrate. It is designed for use as an internal standard or tracer in quantitative analyses and for research on dopamine metabolism and related neurological disorders.
  • Deuterium-labeled internal standard for LC-MS, GC-MS, and NMR.
  • Suitable for quantitative tracer and assay development.
  • Solid hydrate form with stable isotopic labeling.
  • Useful in studies of dopamine metabolism and neurological disease models.
  • Supplied in small research-scale quantities for analytical applications.