Learn More
Medchemexpress LLC 3-O-Methyldopa (Standard) | 300-48-1 | 99.9% | 211.22 | 1 MG
SDP
Catalog No. 5000436831
Encompass_Preferred
3-O-Methyldopa (Standard) is the analytical standard of 3-O-Methyldopa. This product is intended for research and analytical applications. 3-O-Methyldopa (3-Methoxy-L-tyrosine) is a metabolite of L-DOPA, which is formed by catechol-O-methyltransferase (COMT). It competitively inhibits the pharmacodynamics of l-DOPA and dopamine.
  • Analytical standard for qualitative, quantitative, and methodological research experiments
  • Used in HPLC, GC, and MS
Catalog No. 50-004-36831 Supplier Medchemexpress LLC Supplier No. HY113468AR1MG
3-O-Methyldopa (Standard) is the analytical standard of 3-O-Methyldopa. This product is intended for research and analytical applications. 3-O-Methyldopa (3-Methoxy-L-tyrosine) is a metabolite of L-DOPA, which is formed by catechol-O-methyltransferase (COMT). It competitively inhibits the pharmacodynamics of l-DOPA and dopamine.
  • Analytical standard for qualitative, quantitative, and methodological research experiments
  • Used in HPLC, GC, and MS