Learn More
Medchemexpress LLC 3-O-Methyldopa (Standard) | 300-48-1 | 99.9% | 211.22 | 25 MG
SDP
Catalog No. 5000437980
Encompass_Preferred
3-O-Methyldopa (Standard) is the analytical standard of 3-O-Methyldopa. This product is intended for research and analytical applications. 3-O-Methyldopa (3-Methoxy-L-tyrosine) is a metabolite of L-DOPA which is formed by catechol-O-methyltransferase. It competitively inhibits the pharmacodynamics of l-DOPA and dopamine.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in high-performance liquid chromatography, gas chromatography, and mass spectrometry.
Catalog No. 50-004-37980 Supplier Medchemexpress LLC Supplier No. 25MGHY113468AR
3-O-Methyldopa (Standard) is the analytical standard of 3-O-Methyldopa. This product is intended for research and analytical applications. 3-O-Methyldopa (3-Methoxy-L-tyrosine) is a metabolite of L-DOPA which is formed by catechol-O-methyltransferase. It competitively inhibits the pharmacodynamics of l-DOPA and dopamine.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in high-performance liquid chromatography, gas chromatography, and mass spectrometry.