Learn More
Medchemexpress LLC 3-O-Methyldopa (Standard) | 300-48-1 | 99.9% | 211.22 | 50 MG
SDP
Catalog No. 5000437905
Encompass_Preferred
3-O-Methyldopa (Standard) is the analytical standard of 3-O-Methyldopa. It is intended for research and analytical applications. 3-O-Methyldopa (3-Methoxy-L-tyrosine) is a metabolite of L-DOPA, formed by catechol-O-methyltransferase (COMT), and it competitively inhibits the pharmacodynamics of l-DOPA and dopamine.
  • Analytical standard
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: solid
  • Color: white to off-white
  • Initial source: endogenous metabolite
Catalog No. 50-004-37905 Supplier Medchemexpress LLC Supplier No. HY113468AR50MG
3-O-Methyldopa (Standard) is the analytical standard of 3-O-Methyldopa. It is intended for research and analytical applications. 3-O-Methyldopa (3-Methoxy-L-tyrosine) is a metabolite of L-DOPA, formed by catechol-O-methyltransferase (COMT), and it competitively inhibits the pharmacodynamics of l-DOPA and dopamine.
  • Analytical standard
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: solid
  • Color: white to off-white
  • Initial source: endogenous metabolite