Learn More
AdipoGen 3-O-Methylfluorescein
SDP
Catalog No. 502560524
Encompass_Preferred
Chemical. CAS 70672-05-8. Formula C21H14O5. MW 346.33. Synthetic Used in fluorimetric studies. Fluorescent probe. Selective fluorometric substrate for CYP2C19 and CYP1A1. Reaction product of the substrate 3-O-methylfluorescein phosphate OMFP is hydrolyzed by phosphatases to yield O-methylfluorescein OMF that is detected at a 485nm excitation/525nm emission.
Catalog No. 50-256-0524 Supplier AdipoGen Supplier No. CDXM0098M005
Chemical. CAS 70672-05-8. Formula C21H14O5. MW 346.33. Synthetic Used in fluorimetric studies. Fluorescent probe. Selective fluorometric substrate for CYP2C19 and CYP1A1. Reaction product of the substrate 3-O-methylfluorescein phosphate OMFP is hydrolyzed by phosphatases to yield O-methylfluorescein OMF that is detected at a 485nm excitation/525nm emission.