Learn More
Medchemexpress LLC HY-N4167 5mg Medchemexpress, 3-O-Methylgalangin CAS:6665-74-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN41675MG
Encompass_Preferred
Medchemexpress, HY-N4167 5mg 3-O-Methylgalangin CAS:6665-74-3 3-O-Methylgalangin (Galangin 3-methyl ether) is a natural flavonoid compound from the rhizome of Alpinia officinarum (AO) with antibacterial activities, which also inhibits pancreatic lipase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2926